Assay Details
| Gene Symbol: EPCAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002354.2 | NP_002345.2 | 5 - 6 | 914 | 117 | |
| GenBank mRNA | AK026585.1 | - | 5 - 6 | 751 | 117 | |
| BC014785.1 | - | 5 - 6 | 738 | 3861826 | 117 | |
| CR542259.1 | - | 5 - 6 | 556 | 117 | ||
| CR542283.1 | - | 5 - 6 | 556 | 117 | ||
| M26481.1 | - | 5 - 6 | 715 | 117 | ||
| M32306.1 | - | 5 - 6 | 577 | 117 | ||
| M32325.1 | - | 5 - 6 | 710 | 117 | ||
| M33011.1 | - | 5 - 6 | 663 | 117 | ||
| X14758.1 | - | 5 - 6 | 710 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000293.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
ureteric bud development
positive regulation of cell proliferation
signal transduction involved in regulation of gene expression
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
stem cell differentiation
cell-cell adhesion
negative regulation of cell-cell adhesion mediated by cadherin
positive regulation of cell motility
positive regulation of stem cell proliferation
protein binding
protein complex binding
cadherin binding involved in cell-cell adhesion
ureteric bud development
positive regulation of cell proliferation
signal transduction involved in regulation of gene expression
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
stem cell differentiation
cell-cell adhesion
negative regulation of cell-cell adhesion mediated by cadherin
positive regulation of cell motility
positive regulation of stem cell proliferation
protein binding
protein complex binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02004464.hg.1 | ENST00000263735.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004464.hg.1 | ENST00000405271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004464.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02004464.hg.1 | TC02000293.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000419334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000419334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000419334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000419334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009427.hg.1 | ENST00000511599.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200151929.hg.1 | TC0200007506.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000263735.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000405271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000419334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000419334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000419334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000419334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000456133.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000474691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | ENST00000490733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007506.hg.1 | NM_002354.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009427.hg.1 | ENST00000511599.1 | Ensembl |

EPCAM
Genomic Map