Assay Details
| Gene Symbol: USP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020718.3 | NP_065769.3 | 1 - 2 | 628 | 73 | |
| XM_017023496.1 | XP_016878985.1 | 1 - 2 | 659 | 73 | ||
| XM_017023500.1 | XP_016878989.1 | 1 - 2 | 659 | 73 | ||
| GenBank mRNA | AJ586135.2 | - | 1 - 2 | 628 | 73 | |
| AK126752.1 | - | 1 - 2 | 312 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000959.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16013826.hg.1 | ENST00000219689.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013826.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013826.hg.1 | TC16000959.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563129.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013827.hg.1 | ENST00000219689.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013827.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013827.hg.1 | TC16000959.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013828.hg.1 | ENST00000219689.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013828.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16013828.hg.1 | TC16000959.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563129.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000563525.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | ENST00000567975.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009722.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600174875.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600174875.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600174875.hg.1 | TC1600009722.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600174875.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600174875.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600174870.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600174870.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600174870.hg.1 | TC1600009722.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600174870.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600174870.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600174872.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600174872.hg.1 | NM_020718.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600174872.hg.1 | TC1600009722.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600174872.hg.1 | ENST00000219689.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600174872.hg.1 | NM_020718.3 | Refseq |

USP31
Genomic Map