Assay Details
| Gene Symbol: DFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011802.1 | XP_016867291.1 | 1 - 2 | 71 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017011802.1 transcripts.
Set Membership: |
|

DFNA5
Genomic Map