Assay Details
| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001207037.1 | NP_001193966.1 | 3 - 4 | 607 | 76 | |
| GenBank mRNA | AF116508.1 | - | 3 - 4 | 423 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001673.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
organ morphogenesis
cell differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
organ morphogenesis
cell differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000339796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000340181.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373737.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000373738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000538992.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000615781.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000620358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000627426.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000627426.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000627426.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000627426.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000627426.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000627426.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000627426.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000627426.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | ENST00000627426.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207036.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207038.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207039.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_001207041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011658.hg.1 | NM_016135.3 | Refseq |

ETV7
Genomic Map