Assay Details
| Gene Symbol: HSDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146051.1 | NP_001139523.1 | 2 - 3 | 181 | 73 | |
| NM_031463.4 | NP_113651.4 | 2 - 3 | 181 | 73 | ||
| XM_005256189.2 | XP_005256246.1 | 2 - 3 | 149 | 73 | ||
| GenBank mRNA | AK074878.1 | - | 2 - 3 | 145 | 73 | |
| AK299798.1 | - | 2 - 3 | 155 | 73 | ||
| BC018084.1 | - | 2 - 3 | 174 | 4814348 | 73 | |
| BC104218.1 | - | 1 - 2 | 62 | 40032007 | 73 | |
| BC104219.1 | - | 1 - 2 | 62 | 40032008 | 73 | |
| BC106917.2 | - | 1 - 2 | 62 | 40032212 | 73 | |
| DC306668.1 | - | 2 - 3 | 181 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001308.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK074878.1,AK299798.1,BC018084.1,BC104218.1,BC104219.1,BC106917.2,NM_001146051.1,NM_031463.4,XM_005256189.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000565275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000565275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018139.hg.1 | ENST00000219439.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018139.hg.1 | ENST00000434463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018139.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018139.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018139.hg.1 | TC16001308.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000565275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000565275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | ENST00000619773.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011047.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | NM_031463.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | TC1600011047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000219439.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000434463.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000562224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000564998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000567294.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | ENST00000568857.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | NM_001146051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600185821.hg.1 | NM_031463.4 | Refseq |

HSDL1
Genomic Map