| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019302.hg.1 |
ENST00000318967.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019302.hg.1 |
ENST00000435409.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019302.hg.1 |
ENST00000456435.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019302.hg.1 |
ENST00000476843.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019302.hg.1 |
ENST00000512605.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019302.hg.1 |
NM_023015.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01019302.hg.1 |
TC01001263.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000318967.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000435409.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000476843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000481797.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000491790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000503133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000512605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
NM_023015.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000318967.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000435409.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000476843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000481797.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000491790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000503133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
ENST00000512605.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
NM_023015.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100239493.hg.1 |
TC0100018298.hg.1 |
Clariom D TC |