Assay Details
| Gene Symbol: INTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324475.1 | NP_001311404.1 | 26 - 27 | 2785 | 64 | |
| NM_023015.4 | NP_075391.3 | 25 - 26 | 3126 | 64 | ||
| XM_005245459.2 | XP_005245516.1 | 25 - 26 | 3143 | 64 | ||
| XM_005245461.2 | XP_005245518.1 | 19 - 20 | 1816 | 64 | ||
| XM_006711490.3 | XP_006711553.1 | 26 - 27 | 2596 | 64 | ||
| XM_011509906.2 | XP_011508208.1 | 25 - 26 | 2508 | 64 | ||
| XM_017002132.1 | XP_016857621.1 | 15 - 16 | 1384 | 64 | ||
| GenBank mRNA | AK025572.1 | - | 11 - 12 | 947 | 64 | |
| AK055174.1 | - | 5 - 6 | 1311 | 64 | ||
| AK074979.1 | - | 18 - 19 | 1824 | 64 | ||
| AK126799.1 | - | 5 - 6 | 1664 | 64 | ||
| AK225294.1 | - | 11 - 12 | 947 | 64 | ||
| AK290025.1 | - | 26 - 27 | 2779 | 64 | ||
| AK298746.1 | - | 13 - 14 | 1276 | 64 | ||
| AK304874.1 | - | 19 - 20 | 2215 | 64 | ||
| AL080172.1 | - | 20 - 21 | 2025 | 64 | ||
| AL832133.1 | - | 24 - 25 | 4161 | 64 | ||
| BC025254.1 | - | 9 - 10 | 786 | 64 | ||
| BC054513.1 | - | 12 - 13 | 1162 | 64 | ||
| BC073985.1 | - | 9 - 10 | 798 | 64 | ||
| BC098431.1 | - | 11 - 12 | 986 | 64 | ||
| BC105092.1 | - | 25 - 26 | 2589 | 8144095 | 64 | |
| BC105094.1 | - | 25 - 26 | 2589 | 8144097 | 64 | |
| BC116458.1 | - | 25 - 26 | 3121 | 5752962 | 64 | |
| BX640786.1 | - | 25 - 26 | 3112 | 64 | ||
| BX640950.1 | - | 25 - 26 | 3127 | 64 | ||
| CR749212.1 | - | 23 - 24 | 4653 | 64 | ||
| CR749376.1 | - | 24 - 25 | 4778 | 64 | ||
More Information
Set Membership: |
|

INTS3
Genomic Map