Assay Details
| Gene Symbol: EMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011543146.2 | XP_011541448.1 | 1 - 2 | 111 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011543146.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000303221.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000505896.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000506190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000508934.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | ENST00000514111.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010670.hg.1 | NM_198449.2 | Refseq |

EMB
Genomic Map