Assay Details
| Gene Symbol: SSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161330.1 | NP_001154802.1 | 1 - 2 | 160 | 99 | |
| NM_018984.3 | NP_061857.3 | 1 - 2 | 160 | 99 | ||
| GenBank mRNA | AB072355.1 | - | 1 - 2 | 73 | 99 | |
| AB072356.1 | - | 1 - 2 | 73 | 99 | ||
| BC062341.1 | - | 1 - 2 | 152 | 6376684 | 99 | |
| BP363810.1 | - | 1 - 2 | 160 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001941.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
protein dephosphorylation
regulation of actin polymerization or depolymerization
regulation of lamellipodium assembly
actin cytoskeleton organization
regulation of cellular protein metabolic process
peptidyl-tyrosine dephosphorylation
regulation of axonogenesis
cellular response to ATP
DNA binding
actin binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
cell morphogenesis
protein dephosphorylation
regulation of actin polymerization or depolymerization
regulation of lamellipodium assembly
actin cytoskeleton organization
regulation of cellular protein metabolic process
peptidyl-tyrosine dephosphorylation
regulation of axonogenesis
cellular response to ATP
DNA binding
actin binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000326470.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000546812.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000547862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000548522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000548522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000548522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000548522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000548522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000548522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000548522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000548522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000548522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | ENST00000551165.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161330.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_001161331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012841.hg.1 | NM_018984.3 | Refseq |

SSH1
Genomic Map