| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013856.hg.1 |
ENST00000279101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013856.hg.1 |
ENST00000370560.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013856.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013856.hg.1 |
TC20001009.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009748.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168000.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168000.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168000.hg.1 |
NM_031215.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000168000.hg.1 |
TC2000009748.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000168000.hg.1 |
ENST00000279101.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168000.hg.1 |
ENST00000453274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000168000.hg.1 |
NM_031215.2 |
Refseq |