Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003511.hg.1 |
TC04000239.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003511.hg.1 |
ENST00000261435.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003511.hg.1 |
ENST00000381804.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003511.hg.1 |
NM_018177.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149971.hg.1 |
ENST00000261435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149971.hg.1 |
ENST00000511480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149971.hg.1 |
ENST00000513269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149971.hg.1 |
NM_018177.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149971.hg.1 |
ENST00000261435.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149971.hg.1 |
ENST00000511480.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149971.hg.1 |
ENST00000513269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149971.hg.1 |
NM_018177.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149971.hg.1 |
TC0400007306.hg.1 |
Clariom D TC |