| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010338.hg.1 |
ENST00000258052.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010338.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06010338.hg.1 |
TC06000876.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
ENST00000458487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009094.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162508.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162508.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162508.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162508.hg.1 |
ENST00000258052.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162508.hg.1 |
ENST00000439615.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600162508.hg.1 |
NM_003080.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600162508.hg.1 |
TC0600009094.hg.1 |
Clariom D TC |