Assay Details
| Gene Symbol: LSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018385.2 | NP_060855.2 | 7 - 8 | 1080 | 99 | |
| XM_017006796.1 | XP_016862285.1 | 3 - 4 | 408 | 99 | ||
| GenBank mRNA | AK002163.1 | - | 7 - 8 | 813 | 99 | |
| AK225094.1 | - | 7 - 8 | 781 | 99 | ||
| AL136897.1 | - | 7 - 8 | 785 | 99 | ||
| BC015042.1 | - | 7 - 8 | 785 | 99 | ||
| BC040119.1 | - | 7 - 8 | 765 | 99 | ||
| BC058891.1 | - | 7 - 8 | 771 | 99 | ||
| BC068500.1 | - | 7 - 8 | 794 | 30343878 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002137.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03037716.hg.1 | ENST00000265245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037716.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037716.hg.1 | TC03002137.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000461343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000461343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000461343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000461343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037715.hg.1 | ENST00000265245.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037715.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03037715.hg.1 | TC03002137.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300201596.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201596.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201596.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201596.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201596.hg.1 | TC0300013609.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000427461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000460584.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000461343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000461343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000461343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000461343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000466391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000475763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | ENST00000480853.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013609.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201595.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201595.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201595.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201595.hg.1 | ENST00000265245.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201595.hg.1 | ENST00000437613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300201595.hg.1 | NM_018385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300201595.hg.1 | TC0300013609.hg.1 | Clariom D TC |

LSG1
Genomic Map