Assay Details
| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052844.3 | NP_443076.2 | 7 - 8 | 1339 | 59 | |
| XM_011519179.2 | XP_011517481.1 | 7 - 8 | 1189 | 59 | ||
| GenBank mRNA | BC001614.1 | - | 7 - 8 | 1174 | 59 | |
| BC011874.2 | - | 7 - 8 | 1276 | 3161070 | 59 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001634.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09020731.hg.1 | ENST00000372715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020731.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020731.hg.1 | TC09001634.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179669.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179669.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179669.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179669.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179669.hg.1 | TC0900011650.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |

WDR34
Genomic Map