Assay Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052860.2 | NP_443092.1 | 3 - 4 | 436 | 74 | |
| GenBank mRNA | AF395541.1 | - | 3 - 4 | 282 | 74 | |
| AK096465.1 | - | 3 - 4 | 433 | 74 | ||
| BC117248.1 | - | 3 - 4 | 185 | 40125799 | 74 | |
| DA497503.1 | - | 3 - 4 | 437 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF395541.1, AK096465.1, BC117248.1, NM_052860.2 transcripts.
Set Membership: |
|

ZNF300
Genomic Map