Assay Details
| Gene Symbol: C1QBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001212.3 | NP_001203.1 | 2 - 3 | 466 | 72 | |
| GenBank mRNA | AF275902.1 | - | 1 - 2 | 133 | 72 | |
| BC000435.2 | - | 2 - 3 | 420 | 2820039 | 72 | |
| BC013731.1 | - | 2 - 3 | 409 | 3862881 | 72 | |
| BT019898.1 | - | 2 - 3 | 388 | 72 | ||
| BT019899.1 | - | 2 - 3 | 388 | 72 | ||
| L04636.1 | - | 2 - 3 | 466 | 72 | ||
| X75913.1 | - | 2 - 3 | 466 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001056.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
mRNA processing
apoptotic process
immune response
complement activation, classical pathway
blood coagulation, intrinsic pathway
RNA splicing
phosphatidylinositol 3-kinase signaling
viral process
regulation of complement activation
negative regulation of interferon-gamma production
negative regulation of interleukin-12 production
negative regulation of MDA-5 signaling pathway
negative regulation of RIG-I signaling pathway
mature ribosome assembly
positive regulation of apoptotic process
innate immune response
positive regulation of cell adhesion
negative regulation of mRNA splicing, via spliceosome
negative regulation of defense response to virus
positive regulation of protein kinase B signaling
positive regulation of mitochondrial translation
positive regulation of neutrophil chemotaxis
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of trophoblast cell migration
positive regulation of dendritic cell chemotaxis
complement component C1q binding
transcription corepressor activity
mRNA binding
protein kinase C binding
protein binding
hyaluronic acid binding
transcription factor binding
kininogen binding
adrenergic receptor binding
mitochondrial ribosome binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
mRNA processing
apoptotic process
immune response
complement activation, classical pathway
blood coagulation, intrinsic pathway
RNA splicing
phosphatidylinositol 3-kinase signaling
viral process
regulation of complement activation
negative regulation of interferon-gamma production
negative regulation of interleukin-12 production
negative regulation of MDA-5 signaling pathway
negative regulation of RIG-I signaling pathway
mature ribosome assembly
positive regulation of apoptotic process
innate immune response
positive regulation of cell adhesion
negative regulation of mRNA splicing, via spliceosome
negative regulation of defense response to virus
positive regulation of protein kinase B signaling
positive regulation of mitochondrial translation
positive regulation of neutrophil chemotaxis
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of trophoblast cell migration
positive regulation of dendritic cell chemotaxis
complement component C1q binding
transcription corepressor activity
mRNA binding
protein kinase C binding
protein binding
hyaluronic acid binding
transcription factor binding
kininogen binding
adrenergic receptor binding
mitochondrial ribosome binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17014127.hg.1 | ENST00000225698.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014127.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014127.hg.1 | TC17001056.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700204871.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204871.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204871.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204871.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700204871.hg.1 | TC1700012341.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700204871.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204871.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204871.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204871.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700204872.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204872.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204872.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204872.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700204872.hg.1 | TC1700012341.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700204872.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204872.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204872.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700204872.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |

C1QBP
Genomic Map