Assay Details
| Gene Symbol: TMX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005266714.2 | XP_005266771.1 | 4 - 5 | 396 | 127 | |
| GenBank mRNA | AK307269.1 | - | 4 - 5 | 324 | 127 | |
| BC143400.1 | - | 4 - 5 | 366 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005266714.2 transcripts.
Set Membership: |
|

TMX3
Genomic Map