Assay Details
| Gene Symbol: FOXP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244808.1 | NP_001231737.1 | 15 - 16 | 1976 | 87 | |
| XM_005264736.3 | XP_005264793.1 | 10 - 11 | 1611 | 87 | ||
| XM_017006167.1 | XP_016861656.1 | 10 - 11 | 1608 | 87 | ||
| GenBank mRNA | AF275309.1 | - | 6 - 7 | 721 | 87 | |
| AK122710.1 | - | 15 - 16 | 1976 | 87 | ||
| BX647682.1 | - | 9 - 10 | 1063 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

FOXP1
Genomic Map