Assay Details
| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BI910783.1 | - | 1 - 2 | 87 | 70 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |

WDR34
Genomic Map