Assay Details
| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052844.3 | NP_443076.2 | 1 - 2 | 311 | 166 | |
| XM_011519179.2 | XP_011517481.1 | 1 - 2 | 245 | 166 | ||
| GenBank mRNA | BC001614.1 | - | 1 - 2 | 166 | ||
| BC011874.2 | - | 1 - 2 | 3161070 | 166 | ||
| BG177634.1 | - | 1 - 2 | 154 | 165 | ||
| BI910783.1 | - | 2 - 3 | 166 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001634.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020747.hg.1 | ENST00000372715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020747.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020747.hg.1 | TC09001634.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020746.hg.1 | ENST00000372715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020746.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020746.hg.1 | TC09001634.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020749.hg.1 | ENST00000372715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020749.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020749.hg.1 | TC09001634.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020750.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020750.hg.1 | TC09001634.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900179688.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179688.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179688.hg.1 | TC0900011650.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179684.hg.1 | TC0900011650.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000473486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | ENST00000483181.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011650.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179686.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179686.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179686.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179686.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179686.hg.1 | TC0900011650.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | ENST00000419989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | ENST00000451652.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | ENST00000480613.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179683.hg.1 | TC0900011650.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900179687.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179687.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179687.hg.1 | ENST00000372715.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179687.hg.1 | NM_052844.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179687.hg.1 | TC0900011650.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 149145 | - | - |

WDR34
Genomic Map