Assay Details
| Gene Symbol: WDR63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145172.4 | NP_660155.2 | 8 - 9 | 1059 | 124 | |
| GenBank mRNA | AK292603.1 | - | 8 - 9 | 1030 | 124 | |
| AY049724.1 | - | 8 - 9 | 1040 | 124 | ||
| BC040265.1 | - | 8 - 9 | 1013 | 5165779 | 124 | |
| BX648851.1 | - | 8 - 9 | 1065 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

WDR63
Genomic Map