Assay Details
| Gene Symbol: AK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152572.2 | NP_689785.1 | 3 - 4 | 773 | 95 | |
| XM_005272169.2 | XP_005272226.1 | 3 - 4 | 312 | 95 | ||
| XM_011518277.2 | XP_011516579.1 | 3 - 4 | 312 | 95 | ||
| GenBank mRNA | AK057266.1 | - | 3 - 4 | 246 | 95 | |
| AK292186.1 | - | 3 - 4 | 258 | 95 | ||
| BC034776.1 | - | 3 - 4 | 270 | 5175757 | 95 | |
| BC050576.1 | - | 3 - 4 | 773 | 5744517 | 95 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001676.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nucleoside diphosphate phosphorylation
nucleoside triphosphate biosynthetic process
nucleotide biosynthetic process
nucleobase-containing small molecule interconversion
ventricular system development
adenylate kinase activity
cytidylate kinase activity
nucleoside diphosphate kinase activity
protein binding
ATP binding
nucleoside kinase activity
nucleoside diphosphate phosphorylation
nucleoside triphosphate biosynthetic process
nucleotide biosynthetic process
nucleobase-containing small molecule interconversion
ventricular system development
adenylate kinase activity
cytidylate kinase activity
nucleoside diphosphate kinase activity
protein binding
ATP binding
nucleoside kinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000467161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000467161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000467161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000467161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000482422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | ENST00000482422.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021402.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021402.hg.1 | ENST00000477396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021402.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021402.hg.1 | TC09001676.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000467161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000467161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000467161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000467161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000482422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | ENST00000482422.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011815.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900180748.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900180748.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900180748.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900180748.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900180748.hg.1 | ENST00000298545.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900180748.hg.1 | ENST00000476719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900180748.hg.1 | ENST00000477396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900180748.hg.1 | NM_152572.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900180748.hg.1 | TC0900011815.hg.1 | Clariom D TC |

AK8
Genomic Map