| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021391.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021391.hg.1 |
ENST00000477396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021391.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021391.hg.1 |
TC09001676.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000467161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000467161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000467161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000467161.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000482422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
ENST00000482422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000467161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000467161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000467161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000467161.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000482422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
ENST00000482422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011815.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180738.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180738.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180738.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180738.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180738.hg.1 |
ENST00000298545.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180738.hg.1 |
ENST00000476719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180738.hg.1 |
ENST00000477396.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180738.hg.1 |
NM_152572.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180738.hg.1 |
TC0900011815.hg.1 |
Clariom D TC |