Assay Details
| Gene Symbol: SYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003179.2 | NP_003170.1 | 2 - 3 | 120 | 58 | |
| GenBank mRNA | AK313030.1 | - | 2 - 3 | 105 | 58 | |
| BC064550.1 | - | 2 - 3 | 120 | 5768012 | 58 | |
| X06389.1 | - | 1 - 2 | 53 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001029.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
synaptic vesicle maturation
regulation of neuronal synaptic plasticity
regulation of long-term neuronal synaptic plasticity
regulation of short-term neuronal synaptic plasticity
synaptic vesicle membrane organization
cellular response to organic substance
regulation of opioid receptor signaling pathway
transporter activity
cholesterol binding
SH2 domain binding
identical protein binding
protein self-association
endocytosis
synaptic vesicle maturation
regulation of neuronal synaptic plasticity
regulation of long-term neuronal synaptic plasticity
regulation of short-term neuronal synaptic plasticity
synaptic vesicle membrane organization
cellular response to organic substance
regulation of opioid receptor signaling pathway
transporter activity
cholesterol binding
SH2 domain binding
identical protein binding
protein self-association
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013818.hg.1 | ENST00000263233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013818.hg.1 | ENST00000479808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013818.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013818.hg.1 | TC0X001029.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165980.hg.1 | TC0X00009661.hg.1 | Clariom D TC |

SYP
Genomic Map