Assay Details
Gene Symbol: SYP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK295524.1 | - | 2 - 3 | 109 | 61 | |
AK315953.1 | - | 2 - 3 | 109 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK295524.1, AK315953.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
