Assay Details
| Gene Symbol: NUDT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318380.1 | NP_001305309.1 | 3 - 4 | 302 | 99 | |
| NM_177533.4 | NP_803877.2 | 3 - 4 | 302 | 99 | ||
| GenBank mRNA | AB087802.1 | - | 3 - 4 | 188 | 99 | |
| AK128196.1 | - | 3 - 4 | 291 | 99 | ||
| AK310449.1 | - | 3 - 4 | 279 | 99 | ||
| BC041584.1 | - | 3 - 4 | 264 | 4810207 | 99 | |
| DA249638.1 | - | 3 - 4 | 222 | 99 | ||
| DA409636.1 | - | 3 - 4 | 285 | 99 | ||
| DB096906.1 | - | 3 - 4 | 304 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14002334.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017549.hg.1 | ENST00000392568.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017549.hg.1 | ENST00000535832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017549.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017549.hg.1 | TC14002334.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008550.hg.1 | ENST00000392471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000550912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010797.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183531.hg.1 | TC1400010797.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183529.hg.1 | TC1400010797.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000550002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183530.hg.1 | TC1400010797.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | ENST00000339418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | ENST00000392568.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | ENST00000546553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | ENST00000551024.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | NM_177533.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400183532.hg.1 | TC1400010797.hg.1 | Clariom D TC |

NUDT14
Genomic Map