Assay Details
| Gene Symbol: NUDT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105663.2 | NP_001099133.1 | 2 - 3 | 277 | 94 | |
| NM_001243657.1 | NP_001230586.1 | 2 - 3 | 277 | 94 | ||
| XM_011523033.2 | XP_011521335.1 | 2 - 3 | 281 | 94 | ||
| GenBank mRNA | AA227330.1 | - | 2 - 3 | 229 | 94 | |
| BU579534.1 | - | 2 - 3 | 223 | 94 | ||
| BX494638.1 | - | 2 - 3 | 256 | 94 | ||
| CN297379.1 | - | 2 - 3 | 228 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000628.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
nucleoside diphosphate metabolic process
coenzyme A catabolic process
cellular lipid metabolic process
acetyl-CoA catabolic process
brown fat cell differentiation
magnesium ion binding
molecular_function
acetyl-CoA hydrolase activity
receptor binding
CoA hydrolase activity
manganese ion binding
snoRNA binding
m7G(5')pppN diphosphatase activity
biological_process
nucleoside diphosphate metabolic process
coenzyme A catabolic process
cellular lipid metabolic process
acetyl-CoA catabolic process
brown fat cell differentiation
magnesium ion binding
molecular_function
acetyl-CoA hydrolase activity
receptor binding
CoA hydrolase activity
manganese ion binding
snoRNA binding
m7G(5')pppN diphosphatase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16008906.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008906.hg.1 | TC16000628.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000563839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000563839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000563839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000563839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008905.hg.1 | ENST00000268533.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008905.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008905.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008905.hg.1 | TC16000628.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | TC1600008496.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163599.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000268533.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000437314.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000563839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000563839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000563839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000563839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000568787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | ENST00000613075.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001105663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243660.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008496.hg.1 | NM_001243661.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163601.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163601.hg.1 | NM_001243657.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163601.hg.1 | TC1600008496.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600163601.hg.1 | ENST00000564085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163601.hg.1 | NM_001243657.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 159524 | - | - |

NUDT7
Genomic Map