| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
ENST00000261244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
ENST00000354386.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
ENST00000423743.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
ENST00000538571.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
ENST00000556134.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
NM_001244189.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
NM_001244190.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
NM_001244191.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
NM_001244192.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
NM_001244193.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
NM_014749.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14003808.hg.1 |
TC14000351.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000261244.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000354386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000423743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000538571.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000555397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000556134.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000619416.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000619722.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244189.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244190.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244191.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244193.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_014749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000261244.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000354386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000423743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000538571.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000555397.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000556134.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000619416.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
ENST00000619722.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244189.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244190.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244191.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_001244193.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
NM_014749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150775.hg.1 |
TC1400007310.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000261244.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000354386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000423743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000538571.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000556134.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000619416.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000619722.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244189.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244190.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244191.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244193.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_014749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000261244.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000354386.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000423743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000538571.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000556134.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000619416.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
ENST00000619722.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244189.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244190.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244191.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244192.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_001244193.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
NM_014749.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400150776.hg.1 |
TC1400007310.hg.1 |
Clariom D TC |