Assay Details
| Gene Symbol: PPIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005729.3 | NP_005720.1 | 2 - 3 | 314 | 114 | |
| XM_005269379.2 | XP_005269436.1 | 2 - 3 | 312 | 114 | ||
| GenBank mRNA | AK296669.1 | - | 2 - 3 | 307 | 114 | |
| BC005020.1 | - | 2 - 3 | 300 | 3659599 | 114 | |
| BC013204.1 | - | 2 - 3 | 291 | 114 | ||
| BC110299.1 | - | 2 - 3 | 291 | 5215228 | 114 | |
| BE791220.1 | - | 2 - 3 | 330 | 114 | ||
| M80254.1 | - | 2 - 3 | 312 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000566.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein peptidyl-prolyl isomerization
response to ischemia
protein folding
apoptotic mitochondrial changes
regulation of proton-transporting ATPase activity, rotational mechanism
regulation of necrotic cell death
negative regulation of ATPase activity
negative regulation of apoptotic process
regulation of mitochondrial membrane permeability
necroptotic process
cellular response to hydrogen peroxide
cellular response to arsenic-containing substance
cellular response to calcium ion
positive regulation of release of cytochrome c from mitochondria
negative regulation of release of cytochrome c from mitochondria
negative regulation of oxidative phosphorylation
regulation of mitochondrial membrane permeability involved in programmed necrotic cell death
negative regulation of oxidative phosphorylation uncoupler activity
negative regulation of intrinsic apoptotic signaling pathway
peptidyl-prolyl cis-trans isomerase activity
protein binding
cyclosporin A binding
protein peptidyl-prolyl isomerization
response to ischemia
protein folding
apoptotic mitochondrial changes
regulation of proton-transporting ATPase activity, rotational mechanism
regulation of necrotic cell death
negative regulation of ATPase activity
negative regulation of apoptotic process
regulation of mitochondrial membrane permeability
necroptotic process
cellular response to hydrogen peroxide
cellular response to arsenic-containing substance
cellular response to calcium ion
positive regulation of release of cytochrome c from mitochondria
negative regulation of release of cytochrome c from mitochondria
negative regulation of oxidative phosphorylation
regulation of mitochondrial membrane permeability involved in programmed necrotic cell death
negative regulation of oxidative phosphorylation uncoupler activity
negative regulation of intrinsic apoptotic signaling pathway
peptidyl-prolyl cis-trans isomerase activity
protein binding
cyclosporin A binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10006109.hg.1 | ENST00000225174.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006109.hg.1 | ENST00000394579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006109.hg.1 | ENST00000472580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006109.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006109.hg.1 | TC10000566.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006107.hg.1 | ENST00000225174.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006107.hg.1 | ENST00000394579.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006107.hg.1 | ENST00000472580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006107.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006107.hg.1 | TC10000566.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155462.hg.1 | TC1000008193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155460.hg.1 | TC1000008193.hg.1 | Clariom D TC |

PPIF
Genomic Map