| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005968.hg.1 |
ENST00000374688.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005968.hg.1 |
ENST00000374692.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005968.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09005968.hg.1 |
TC09000539.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900155786.hg.1 |
TC0900008318.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374688.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000374692.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000434214.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000435034.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
ENST00000451560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900008318.hg.1 |
NM_018112.2 |
Refseq |