| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020618.hg.1 |
ENST00000296679.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020618.hg.1 |
ENST00000414719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020618.hg.1 |
ENST00000507029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020618.hg.1 |
NM_018268.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020618.hg.1 |
TC05001507.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020621.hg.1 |
ENST00000296679.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020621.hg.1 |
ENST00000414719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020621.hg.1 |
ENST00000507029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020621.hg.1 |
NM_018268.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020621.hg.1 |
TC05001507.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020612.hg.1 |
ENST00000296679.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020612.hg.1 |
ENST00000414719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020612.hg.1 |
ENST00000507029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020612.hg.1 |
NM_018268.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020612.hg.1 |
TC05001507.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020619.hg.1 |
ENST00000296679.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020619.hg.1 |
ENST00000414719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020619.hg.1 |
ENST00000507029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020619.hg.1 |
NM_018268.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020619.hg.1 |
TC05001507.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020620.hg.1 |
ENST00000296679.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020620.hg.1 |
ENST00000414719.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020620.hg.1 |
ENST00000507029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020620.hg.1 |
NM_018268.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05020620.hg.1 |
TC05001507.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000296679.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000414719.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000503035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000507029.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000507452.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000509701.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000509892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000511036.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000513417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000514559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000515253.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000515321.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
NM_018268.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000296679.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000414719.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000503035.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000507029.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000507452.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000509701.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000509892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000511036.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000513417.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000514559.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000515253.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
ENST00000515321.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
NM_018268.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177729.hg.1 |
TC0500011208.hg.1 |
Clariom D TC |