Assay Details
| Gene Symbol: C16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320540.1 | NP_001307469.1 | 13 - 14 | 1141 | 67 | |
| NM_001320541.1 | NP_001307470.1 | 13 - 14 | 1182 | 67 | ||
| NM_001320542.1 | NP_001307471.1 | 13 - 14 | 1167 | 67 | ||
| NM_001320543.1 | NP_001307472.1 | 12 - 13 | 1098 | 67 | ||
| NM_025187.4 | NP_079463.2 | 12 - 13 | 1197 | 67 | ||
| XM_006721284.3 | XP_006721347.1 | 12 - 13 | 1456 | 67 | ||
| XM_011523350.2 | XP_011521652.1 | 11 - 12 | 1596 | 67 | ||
| XM_017023729.1 | XP_016879218.1 | 12 - 13 | 1467 | 67 | ||
| GenBank mRNA | AF176088.1 | - | 13 - 14 | 1097 | 67 | |
| BC004556.2 | - | 12 - 13 | 1177 | 3957147 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000529.hg.1
Set Membership: |
|

C16orf70
Genomic Map