Assay Details
| Gene Symbol: ARV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022786.1 | NP_073623.1 | 4 - 5 | 697 | 96 | |
| XM_017002106.1 | XP_016857595.1 | 5 - 6 | 791 | 96 | ||
| GenBank mRNA | AF271780.1 | - | 4 - 5 | 697 | 96 | |
| AF290878.1 | - | 4 - 5 | 697 | 96 | ||
| AF321442.1 | - | 4 - 5 | 731 | 96 | ||
| AK293049.1 | - | 4 - 5 | 703 | 96 | ||
| AL122047.1 | - | 1 - 2 | 96 | 96 | ||
| BC016309.1 | - | 4 - 5 | 692 | 4047121 | 96 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001913.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
sphingolipid metabolic process
cholesterol biosynthetic process
bile acid metabolic process
sterol metabolic process
cholesterol transport
intracellular sterol transport
regulation of cholesterol metabolic process
regulation of plasma membrane sterol distribution
sterol transporter activity
sphingolipid metabolic process
cholesterol biosynthetic process
bile acid metabolic process
sterol metabolic process
cholesterol transport
intracellular sterol transport
regulation of cholesterol metabolic process
regulation of plasma membrane sterol distribution
sterol transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01029733.hg.1 | ENST00000310256.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029733.hg.1 | ENST00000366658.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029733.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029733.hg.1 | TC01001913.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029732.hg.1 | ENST00000310256.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029732.hg.1 | ENST00000366658.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029732.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029732.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01029732.hg.1 | TC01001913.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188168.hg.1 | TC0100011953.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188169.hg.1 | TC0100011953.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000310256.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000366658.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000435927.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000450711.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000459891.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000480519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | ENST00000497753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | NM_022786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011953.hg.1 | NM_022786.2 | Refseq |

ARV1
Genomic Map