| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024882.hg.1 |
ENST00000335266.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024882.hg.1 |
ENST00000358577.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024882.hg.1 |
ENST00000358807.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024882.hg.1 |
ENST00000368952.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024882.hg.1 |
ENST00000368957.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024882.hg.1 |
NM_001159291.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024882.hg.1 |
NM_022765.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06024882.hg.1 |
TC06002003.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000358577.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000358807.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000456101.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000465904.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000630715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
NM_001159291.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
NM_001286613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
NM_022765.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000358577.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000358807.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000456101.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000465904.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
ENST00000630715.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
NM_001159291.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
NM_001286613.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
NM_022765.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600202466.hg.1 |
TC0600014333.hg.1 |
Clariom D TC |