| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041984.hg.1 |
ENST00000234725.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041984.hg.1 |
ENST00000360494.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041984.hg.1 |
ENST00000371429.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041984.hg.1 |
ENST00000537333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041984.hg.1 |
ENST00000540001.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041984.hg.1 |
NM_001168551.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041984.hg.1 |
NM_018087.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041984.hg.1 |
NR_033142.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01041984.hg.1 |
TC01002676.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100206632.hg.1 |
ENST00000371429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206632.hg.1 |
NM_001168551.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206632.hg.1 |
NM_018087.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206632.hg.1 |
NR_033142.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206632.hg.1 |
ENST00000371429.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100206632.hg.1 |
NM_001168551.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206632.hg.1 |
NM_018087.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206632.hg.1 |
NR_033142.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100206632.hg.1 |
TC0100014243.hg.1 |
Clariom D TC |