Assay Details
| Gene Symbol: UPF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080632.2 | NP_542199.1 | 7 - 8 | 906 | 120 | |
| XM_017029737.1 | XP_016885226.1 | 7 - 8 | 906 | 120 | ||
| XM_017029738.1 | XP_016885227.1 | 7 - 8 | 906 | 120 | ||
| XM_017029740.1 | XP_016885229.1 | 7 - 8 | 906 | 120 | ||
| GenBank mRNA | AF318576.1 | - | 7 - 8 | 826 | 120 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001315.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
mRNA 3'-end processing
positive regulation of translation
nucleotide binding
mRNA binding
nucleocytoplasmic transporter activity
protein binding
poly(A) RNA binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
mRNA 3'-end processing
positive regulation of translation
nucleotide binding
mRNA binding
nucleocytoplasmic transporter activity
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017675.hg.1 | ENST00000276201.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017675.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017675.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X017675.hg.1 | TC0X001315.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00172378.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00172378.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00172378.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00172378.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00172378.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00172378.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00172378.hg.1 | TC0X00010652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000276201.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000345865.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000478840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | ENST00000619445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_023010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010652.hg.1 | NM_080632.2 | Refseq |

UPF3B
Genomic Map