Assay Details
| Gene Symbol: PPIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK296669.1 | - | 4 - 5 | 490 | 66 | |
| BC110299.1 | - | 4 - 5 | 474 | 5215228 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155467.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155467.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155467.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155467.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155467.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155467.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155467.hg.1 | TC1000008193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000155468.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155468.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155468.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155468.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155468.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155468.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155468.hg.1 | TC1000008193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000155470.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155470.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155470.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155470.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155470.hg.1 | TC1000008193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000448165.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000472580.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000492149.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | ENST00000498681.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008193.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155469.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155469.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155469.hg.1 | ENST00000225174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000155469.hg.1 | NM_005729.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000155469.hg.1 | TC1000008193.hg.1 | Clariom D TC |

PPIF
Genomic Map