| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19000200.hg.1 |
ENST00000263620.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19000200.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19000200.hg.1 |
TC19000023.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000592216.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
ENST00000592216.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000585895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000590749.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000592216.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
ENST00000592216.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006499.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
TC1900006499.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
ENST00000585733.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144612.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900144611.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144611.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144611.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144611.hg.1 |
NM_005224.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900144611.hg.1 |
TC1900006499.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900144611.hg.1 |
ENST00000263620.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144611.hg.1 |
ENST00000457152.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144611.hg.1 |
ENST00000587532.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900144611.hg.1 |
NM_005224.2 |
Refseq |