Assay Details
| Gene Symbol: HIF1AN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017902.2 | NP_060372.2 | 1 - 2 | 251 | 74 | |
| GenBank mRNA | AF395830.1 | - | 1 - 2 | 186 | 74 | |
| AK000622.1 | - | 1 - 2 | 196 | 74 | ||
| AK295353.1 | - | 1 - 2 | 278 | 74 | ||
| BC007719.2 | - | 1 - 2 | 179 | 4138066 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000724.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
peptidyl-histidine hydroxylation
peptidyl-aspartic acid hydroxylation
peptidyl-asparagine hydroxylation
positive regulation of myoblast differentiation
negative regulation of Notch signaling pathway
oxidation-reduction process
regulation of transcription from RNA polymerase II promoter in response to hypoxia
negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
positive regulation of vasculogenesis
Notch binding
iron ion binding
protein binding
zinc ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
oxygen sensor activity
carboxylic acid binding
peptidyl-histidine dioxygenase activity
peptidyl-asparagine 3-dioxygenase activity
protein homodimerization activity
cofactor binding
NF-kappaB binding
ankyrin repeat binding
transcription, DNA-templated
peptidyl-histidine hydroxylation
peptidyl-aspartic acid hydroxylation
peptidyl-asparagine hydroxylation
positive regulation of myoblast differentiation
negative regulation of Notch signaling pathway
oxidation-reduction process
regulation of transcription from RNA polymerase II promoter in response to hypoxia
negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
positive regulation of vasculogenesis
Notch binding
iron ion binding
protein binding
zinc ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
oxygen sensor activity
carboxylic acid binding
peptidyl-histidine dioxygenase activity
peptidyl-asparagine 3-dioxygenase activity
protein homodimerization activity
cofactor binding
NF-kappaB binding
ankyrin repeat binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10008424.hg.1 | ENST00000299163.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008424.hg.1 | ENST00000442724.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008424.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008424.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008424.hg.1 | TC10000724.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008422.hg.1 | ENST00000299163.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008422.hg.1 | ENST00000442724.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008422.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008422.hg.1 | TC10000724.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008425.hg.1 | ENST00000299163.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008425.hg.1 | ENST00000442724.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008425.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008425.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008425.hg.1 | TC10000724.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008420.hg.1 | ENST00000299163.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008420.hg.1 | ENST00000442724.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008420.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10008420.hg.1 | TC10000724.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000158997.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158997.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158997.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158997.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158997.hg.1 | TC1000008651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158996.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158996.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158996.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158996.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158996.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158996.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158996.hg.1 | TC1000008651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158999.hg.1 | TC1000008651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000159000.hg.1 | TC1000008651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000478787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000528044.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | ENST00000533589.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008651.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158994.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158994.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158994.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158994.hg.1 | ENST00000299163.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158994.hg.1 | ENST00000526476.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000158994.hg.1 | NM_017902.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000158994.hg.1 | TC1000008651.hg.1 | Clariom D TC |

HIF1AN
Genomic Map