| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000528044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000528044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000528044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000528044.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008431.hg.1 |
ENST00000299163.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008431.hg.1 |
ENST00000442724.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008431.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10008431.hg.1 |
TC10000724.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000478787.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000526476.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000528044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000528044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000528044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000528044.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008651.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159007.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159007.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159007.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159007.hg.1 |
ENST00000299163.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159007.hg.1 |
ENST00000533589.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000159007.hg.1 |
NM_017902.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000159007.hg.1 |
TC1000008651.hg.1 |
Clariom D TC |