Assay Details
| Gene Symbol: EMILIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032048.2 | NP_114437.2 | 4 - 5 | 2520 | 110 | |
| GenBank mRNA | AF270513.1 | - | 4 - 5 | 2520 | 110 | |
| AK090519.1 | - | 1 - 2 | 615 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000014.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18000164.hg.1 | ENST00000254528.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000164.hg.1 | ENST00000308080.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000164.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000164.hg.1 | TC18000014.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000163.hg.1 | ENST00000254528.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000163.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000163.hg.1 | TC18000014.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144496.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144496.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144496.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800144496.hg.1 | TC1800006493.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800144496.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144496.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800144496.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000254528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000308080.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | ENST00000583776.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006493.hg.1 | NM_032048.2 | Refseq |

EMILIN2
Genomic Map