Assay Details
| Gene Symbol: GLRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042543.2 | NP_001036008.1 | 5 - 6 | 1078 | 118 | |
| NM_006529.3 | NP_006520.2 | 5 - 6 | 1078 | 118 | ||
| XM_011532267.2 | XP_011530569.1 | 5 - 6 | 1331 | 118 | ||
| XM_017008630.1 | XP_016864119.1 | 6 - 7 | 1102 | 118 | ||
| GenBank mRNA | BC036086.1 | - | 5 - 6 | 1077 | 5286731 | 118 |
| U93917.1 | - | 5 - 6 | 994 | 118 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001755.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neuropeptide signaling pathway
ion transmembrane transport
response to amino acid
protein homooligomerization
synaptic transmission, glycinergic
chloride transmembrane transport
glycine binding
extracellular-glycine-gated chloride channel activity
glycine-gated chloride ion channel activity
metal ion binding
neuropeptide signaling pathway
ion transmembrane transport
response to amino acid
protein homooligomerization
synaptic transmission, glycinergic
chloride transmembrane transport
glycine binding
extracellular-glycine-gated chloride channel activity
glycine-gated chloride ion channel activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04024204.hg.1 | ENST00000274093.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024204.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024204.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024204.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024204.hg.1 | TC04001755.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000436738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | ENST00000436738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000436738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | ENST00000436738.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012462.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400185449.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400185449.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400185449.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400185449.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400185449.hg.1 | ENST00000274093.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400185449.hg.1 | ENST00000340217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400185449.hg.1 | NM_001042543.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400185449.hg.1 | NM_006529.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400185449.hg.1 | TC0400012462.hg.1 | Clariom D TC |

GLRA3
Genomic Map