Assay Details
| Gene Symbol: SNRPGP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BF085802.1 | - | 1 | 44 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3263170 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3212679 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 23814 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12220 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3263170 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3212679 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892526 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892519 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892529 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509214 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892524 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892523 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112452 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509224 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892537 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892521 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892533 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892536 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892522 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892530 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112444 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892525 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892520 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892532 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892528 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892518 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112443 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509216 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112455 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112447 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509222 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509223 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509228 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892527 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112445 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892535 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509209 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892534 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509212 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892516 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892517 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509225 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509203 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892531 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509211 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892532 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892531 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892533 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892523 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112452 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112445 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509228 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509214 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509228 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892530 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892529 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892527 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892535 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509209 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892536 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509211 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892524 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112447 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892533 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112455 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892526 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509216 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509222 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112451 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892519 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892531 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892521 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509224 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112443 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892516 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892518 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892537 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892525 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892534 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112452 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892532 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892523 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509203 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892520 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509225 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509212 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892517 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892522 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31509223 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112444 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10892528 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17112445 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112445 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112447 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509222 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509214 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892516 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892532 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509224 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892533 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892537 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892531 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509209 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112452 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892522 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10892523 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17112455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31509223 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112452 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112451 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892520 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892562 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892552 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892537 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892550 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892532 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892531 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892535 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892528 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892516 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112469 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892519 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112455 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112465 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892561 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892526 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892544 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112454 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892548 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892551 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892546 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10892554 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112447 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112464 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112449 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17112470 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8026340 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8026339 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A012309 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A012309 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 170316 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3258321 | NM_001146693.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3192156 | NM_001146693.1 | human_transcriptome |

SNRPGP15
Genomic Map