Assay Details
| Gene Symbol: CTSD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001909.4 | NP_001900.1 | 4 - 5 | 603 | 64 | |
| GenBank mRNA | AK130178.1 | - | 4 - 5 | 495 | 64 | |
| BC016320.2 | - | 4 - 5 | 515 | 3506977 | 64 | |
| BT006910.1 | - | 4 - 5 | 470 | 64 | ||
| BT020155.1 | - | 4 - 5 | 470 | 64 | ||
| CR456947.1 | - | 4 - 5 | 470 | 64 | ||
| EU794598.1 | - | 4 - 5 | 515 | 64 | ||
| KU178007.1 | - | 4 - 5 | 470 | 64 | ||
| M11233.1 | - | 4 - 5 | 521 | 64 | ||
| X05344.1 | - | 4 - 5 | 472 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003472.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
autophagy
antigen processing and presentation of exogenous peptide antigen via MHC class II
protein catabolic process
collagen catabolic process
aspartic-type endopeptidase activity
cysteine-type endopeptidase activity
serine-type endopeptidase activity
protein binding
proteolysis
autophagy
antigen processing and presentation of exogenous peptide antigen via MHC class II
protein catabolic process
collagen catabolic process
aspartic-type endopeptidase activity
cysteine-type endopeptidase activity
serine-type endopeptidase activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11016035.hg.1 | ENST00000236671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016035.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016035.hg.1 | TC11003472.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000429746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | ENST00000497544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013129.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | ENST00000236671.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | ENST00000367196.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | ENST00000433655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | ENST00000438213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | NM_001909.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203289.hg.1 | TC1100013129.hg.1 | Clariom D TC |

CTSD
Genomic Map