Assay Details
| Gene Symbol: GTPBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014170.2 | NP_054889.2 | 2 - 3 | 483 | 97 | |
| GenBank mRNA | AF161484.1 | - | 2 - 3 | 95 | ||
| AK093419.1 | - | 2 - 3 | 1077 | 97 | ||
| AK292651.1 | - | 2 - 3 | 502 | 97 | ||
| BC037163.1 | - | 2 - 3 | 486 | 4620206 | 97 | |
| BI830308.1 | - | 2 - 3 | 483 | 97 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000575.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010456.hg.1 | ENST00000305485.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010456.hg.1 | ENST00000383677.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010456.hg.1 | ENST00000383678.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010456.hg.1 | ENST00000467752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010456.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010456.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010456.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03010456.hg.1 | TC03000575.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000295864.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000488781.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008352.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000383677.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000383678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000467752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000473129.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000485330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000618381.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | ENST00000619116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | NM_014170.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | NM_138485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300159558.hg.1 | TC0300008352.hg.1 | Clariom D TC |

GTPBP8
Genomic Map