Assay Details
| Gene Symbol: GJB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006783.4 | NP_006774.2 | 1 - 2 | 374 | 74 | |
| GenBank mRNA | AK289592.1 | - | 1 - 2 | 122 | 74 | |
| AK312352.1 | - | 1 - 2 | 80 | 74 | ||
| BC038934.1 | - | 1 - 2 | 69 | 5196769 | 74 | |
| DC337465.1 | - | 1 - 2 | 374 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000457.hg
Additional Information:
This assay is in the 5'UTR region of the AK289592.1,BC038934.1,NM_006783.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13005202.hg.1 | ENST00000241124.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005202.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005202.hg.1 | ENST00000400065.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005202.hg.1 | ENST00000400066.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005202.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005202.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005202.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005202.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005202.hg.1 | TC13000457.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005206.hg.1 | ENST00000241124.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005206.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005206.hg.1 | TC13000457.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005207.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005207.hg.1 | TC13000457.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005205.hg.1 | ENST00000241124.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005205.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005205.hg.1 | TC13000457.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300155682.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155682.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155682.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155682.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155682.hg.1 | TC1300008250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300155681.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155681.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155681.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155681.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155681.hg.1 | TC1300008250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300155683.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155683.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155683.hg.1 | TC1300008250.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008250.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | ENST00000241124.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | ENST00000356192.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | ENST00000400065.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | ENST00000400066.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | NM_001110219.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | NM_001110220.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | NM_001110221.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | NM_006783.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300155677.hg.1 | TC1300008250.hg.1 | Clariom D TC |

GJB6
Genomic Map