| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015465.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015465.hg.1 |
ENST00000377041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015465.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015465.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015465.hg.1 |
TC09001205.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015467.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015467.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015467.hg.1 |
TC09001205.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015468.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015468.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015468.hg.1 |
TC09001205.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015464.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015464.hg.1 |
ENST00000377041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015464.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015464.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015464.hg.1 |
TC09001205.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015466.hg.1 |
ENST00000377041.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015466.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015466.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09015466.hg.1 |
TC09001205.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010390.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171114.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171114.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171114.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171114.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171114.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171114.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171114.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171114.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171114.hg.1 |
TC0900010390.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900171116.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171116.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171116.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171116.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171116.hg.1 |
ENST00000333421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171116.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171116.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171116.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171116.hg.1 |
TC0900010390.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900171117.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171117.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171117.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171117.hg.1 |
ENST00000377041.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900171117.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171117.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171117.hg.1 |
TC0900010390.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900171118.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171118.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171118.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171118.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171118.hg.1 |
TC0900010390.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900171119.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171119.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171119.hg.1 |
NM_001025780.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171119.hg.1 |
NM_016014.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900171119.hg.1 |
TC0900010390.hg.1 |
Clariom D TC |