| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001981.hg.1 |
ENST00000312541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001981.hg.1 |
ENST00000429381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001981.hg.1 |
ENST00000469185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001981.hg.1 |
ENST00000541522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001981.hg.1 |
NM_001197233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001981.hg.1 |
NM_001197234.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001981.hg.1 |
NM_007049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001981.hg.1 |
NM_078476.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001981.hg.1 |
TC06000189.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
ENST00000265424.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
ENST00000312541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
ENST00000429381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
ENST00000469185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
ENST00000541522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
NM_001197233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
NM_001197234.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
NM_007049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
NM_078476.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001979.hg.1 |
TC06000189.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
ENST00000265424.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
ENST00000312541.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
ENST00000429381.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
ENST00000469185.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
ENST00000541522.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
NM_001197233.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
NM_001197234.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
NM_007049.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
NM_078476.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06001982.hg.1 |
TC06000189.hg.1 |
HTA TC |