Assay Details
| Gene Symbol: CERK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022766.5 | NP_073603.2 | 2 - 3 | 362 | 68 | |
| XM_017028909.1 | XP_016884398.1 | 2 - 3 | 159 | 68 | ||
| GenBank mRNA | AB051433.1 | - | 1 - 2 | 88 | 68 | |
| AB079066.1 | - | 2 - 3 | 378 | 68 | ||
| AJ457828.1 | - | 2 - 3 | 325 | 68 | ||
| AK291476.1 | - | 2 - 3 | 318 | 68 | ||
| BC067255.1 | - | 2 - 3 | 314 | 68 | ||
| BC126940.1 | - | 2 - 3 | 305 | 40084110 | 68 | |
| CR456404.1 | - | 2 - 3 | 265 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000890.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC067255.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22014982.hg.1 | ENST00000216264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014982.hg.1 | ENST00000541677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014982.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014982.hg.1 | TC22000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014984.hg.1 | ENST00000216264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014984.hg.1 | ENST00000541677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014984.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014984.hg.1 | TC22000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014985.hg.1 | ENST00000216264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014985.hg.1 | ENST00000541677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014985.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014985.hg.1 | TC22000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014981.hg.1 | ENST00000216264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014981.hg.1 | ENST00000541677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014981.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014981.hg.1 | TC22000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014987.hg.1 | ENST00000216264.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014987.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22014987.hg.1 | TC22000890.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165427.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165427.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165427.hg.1 | TC2200009028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165427.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165427.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165421.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165421.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165421.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165421.hg.1 | TC2200009028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165421.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165421.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165421.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165425.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165425.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165425.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165425.hg.1 | TC2200009028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165425.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165425.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165425.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165420.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165420.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165420.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165420.hg.1 | TC2200009028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165420.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165420.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165420.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165423.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165423.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165423.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165423.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165423.hg.1 | TC2200009028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165423.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165423.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165423.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165423.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165424.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165424.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165424.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200165424.hg.1 | TC2200009028.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200165424.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165424.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200165424.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |

CERK
Genomic Map