Assay Details
Gene Symbol: CERK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022766.5 | NP_073603.2 | 9 - 10 | 1157 | 71 | |
XM_017028909.1 | XP_016884398.1 | 9 - 10 | 954 | 71 | ||
GenBank mRNA | AB051433.1 | - | 8 - 9 | 883 | 71 | |
AB079066.1 | - | 9 - 10 | 1173 | 71 | ||
AJ457828.1 | - | 9 - 10 | 1120 | 71 | ||
AK291476.1 | - | 9 - 10 | 1113 | 71 | ||
BC067255.1 | - | 8 - 9 | 1034 | 71 | ||
BC126940.1 | - | 9 - 10 | 1100 | 40084110 | 71 | |
CR456404.1 | - | 9 - 10 | 1060 | 71 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000890.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22014973.hg.1 | ENST00000216264.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014973.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014973.hg.1 | ENST00000541677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014973.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22014973.hg.1 | TC22000890.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22014972.hg.1 | ENST00000216264.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014972.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014972.hg.1 | ENST00000541677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014972.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22014972.hg.1 | TC22000890.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000460254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009028.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165411.hg.1 | TC2200009028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200165411.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165411.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165411.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165411.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165411.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165411.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165411.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165411.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165412.hg.1 | TC2200009028.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200165412.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165412.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165412.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165412.hg.1 | NM_022766.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200165412.hg.1 | ENST00000216264.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165412.hg.1 | ENST00000443629.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165412.hg.1 | ENST00000471929.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200165412.hg.1 | NM_022766.5 | Refseq |
